missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Human CAMK2B (CaMKII Beta), His Tag Recombinant Protein
Human CAMK2B (CaMKII Beta), His Tag Recombinant Protein
5380.00 SEK - 18260.00 SEK
Specifications
Accession Number | NP_742078 |
---|---|
For Use With (Application) | Kinase Assay |
Molecular Weight (g/mol) | 60.5 kDa |
Gene Alias | CAMK2B (CaMKII Beta), CAMK2 |
KinaseFamily | CAMK2 Kinase Family |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15448623
|
Thermo Scientific™
PR7771A |
100 μg |
18260.00 SEK
100µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11573627
|
Thermo Scientific™
PV4205 |
10 μg |
5380.00 SEK
10µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
As a dominant kinase in the central nervous system CAMK2B (CaMKII beta) may function in long term potentiation and neurotransmitter release.Specifications
NP_742078 | |
60.5 kDa | |
CAMK2 Kinase Family | |
Recombinant | |
Storage Buffer: 50 mM Tris pH 7.5, 150 mM NaCl, 0.5 mM EDTA, 0.02% Triton X-100, 2 mM DTT, and 50% Glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Dry Ice | |
Kinases & Inhibitors |
Kinase Assay | |
CAMK2B (CaMKII Beta), CAMK2 | |
Human | |
His Tag | |
Ser⁄Thr Kinases: CAMK Group | |
Baculovirus |