Learn More
ACROBiosystems Canine TNF-alpha Protein, His Tag
Brand:  ACROBiosystems TNA-C52H3-100ug
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 17216590
  • 4673.08 SEK / 100µg
Estimated Shipment: 05-06-2024
to see stock.
Description
Description
Tumor necrosis factor alpha (TNFα) is a cytokine produced primarily by monocytes and macrophages. It is found in synovial cells and macrophages in the tissues.The primary role of TNFα is in the regulation of immune cells. TNFα is able to induce apoptotic cell death, to induce inflammation, and to inhibit tumorigenesis and viral replication. Dysregulation of TNFα production has been implicated in a variety of human diseases, including major depression, Alzheimer's disease and cancer. Recombinant TNFα is used as an immunostimulant under the INN tasonermin. TNFα can be produced ectopically in the setting of malignancy and parallels parathyroid hormone both in causing secondary hypercalcemia and in the cancers with which excessive production is associated.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 19.2 kDa.
Specifications
Specifications
19.2 kDa
Canine TNF-alpha, His Tag (TNA-C52H3) is expressed from human 293 cells (HEK293). It contains AA Val 77 - Leu 233 (Accession # P51742-1).
-20°C
TNF-alpha
Canine
HEK293
Cytokines&Growth; Factors
100 μg
Val 77 - Leu 233
DIF,TNF-alpha,TNFA,TNFSF2,cachexin,cachectin,TNFα
Unconjugated
His Tag
Powder