missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cannabinoid R1/CB1/CNR1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Bio-Techne NB300-552
This item is not returnable.
View return policy
Description
Cannabinoid R1/CB1/CNR1 Polyclonal specifically detects Cannabinoid R1/CB1/CNR1 in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
Cannabinoid R1/CB1/CNR1 | |
Polyclonal | |
Unconjugated | |
PBS, 1.0 mg/ml BSA and 30% Glycerol with 0.05% Sodium Azide | |
CNR1 | |
A fusion protein containing the first 99 amino acid residues from human CB1. | |
100 ÎĽL | |
Cancer, GPCR, Neuroscience, Neurotransmission, Vision | |
1268 | |
Human, Rat | |
IgG |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
1 mg/mL | |
Western Blot 1:250, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:1000, Immunohistochemistry-Paraffin 1:10 - 1:500, Immunohistochemistry-Frozen 1:10 - 1:500 | |
CANN6CB1R, cannabinoid receptor 1 (brain), CB1A, CB1K5, CB-RCB1cannabinoid receptor 1, central cannabinoid receptor, CNR | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Detects CB1 from human and rat tissues as well as transfected rat CB1. | |
Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Cannabinoid R1/CB1/CNR1 Antibody, Novus Biologicals™ > 0.1mL; Unlabeled
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction