Läs mer
Canrenoic acid potassium salt, 98+%, Thermo Scientific™
Brand: Thermo Scientific Alfa Aesar J61394.06
| Kvantitet | 5g |
|---|
Beskrivning
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Kemiska identifierare
| 2181-04-6 | |
| 396.57 | |
| JTZQCHFUGHIPDF-RYVBEKKQSA-M | |
| potassium;3-[(10S,13R,17S)-17-hydroxy-10,13-dimethyl-3-oxo-2,8,9,11,12,14,15,16-octahydro-1H-cyclopenta[a]phenanthren-17-yl]propanoate |
| C22H29KO4 | |
| MFCD05662375 | |
| 133109093 | |
| CC12CCC(=O)C=C1C=CC3C2CCC4(C3CCC4(CCC(=O)[O-])O)C.[K+] |
Specifikationer
| Canrenoic acid potassium salt | |
| 5g | |
| MFCD05662375 | |
| JTZQCHFUGHIPDF-RYVBEKKQSA-M | |
| potassium;3-[(10S,13R,17S)-17-hydroxy-10,13-dimethyl-3-oxo-2,8,9,11,12,14,15,16-octahydro-1H-cyclopenta[a]phenanthren-17-yl]propanoate | |
| 133109093 | |
| ≥98% |
| 2181-04-6 | |
| C22H29KO4 | |
| 14,1750 | |
| CC12CCC(=O)C=C1C=CC3C2CCC4(C3CCC4(CCC(=O)[O-])O)C.[K+] | |
| 396.57 | |
| 396.58 |