Läs mer
Canrenone, 98%, Thermo Scientific™
Inhibits aldosterone biosynthesis and blocks ouabain effects
Brand: Thermo Scientific Alfa Aesar J60238.14
| Kvantitet | 25g |
|---|
Beskrivning
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Kemiska identifierare
| C22H28O3 | |
| MFCD00071735 | |
| canrenone, aldadiene, phanurane, luvion, aldadiene van, canrenone usan:inn, canrenonum inn-latin, canrenona inn-spanish, unii-78o20x9j0u, 17alpha-2-carboxyethyl-17beta-hydroxyandrosta-4,6-dien-3-one lactone | |
| (8R,9S,10R,13S,14S,17R)-10,13-dimethylspiro[2,8,9,11,12,14,15,16-octahydro-1H-cyclopenta[a]phenanthrene-17,5'-oxolane]-2',3-dione |
Specifikationer
| Canrenone | |
| C22H28O3 | |
| 25g | |
| canrenone, aldadiene, phanurane, luvion, aldadiene van, canrenone usan:inn, canrenonum inn-latin, canrenona inn-spanish, unii-78o20x9j0u, 17alpha-2-carboxyethyl-17beta-hydroxyandrosta-4,6-dien-3-one lactone | |
| CC12CCC(=O)C=C1C=CC3C2CCC4(C3CCC45CCC(=O)O5)C | |
| 340.463 | |
| 340.46 |
| 976-71-6 | |
| MFCD00071735 | |
| 14,1752 | |
| UJVLDDZCTMKXJK-WNHSNXHDSA-N | |
| (8R,9S,10R,13S,14S,17R)-10,13-dimethylspiro[2,8,9,11,12,14,15,16-octahydro-1H-cyclopenta[a]phenanthrene-17,5'-oxolane]-2',3-dione | |
| 13789 | |
| 98% |