Learn More
CAP2 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15865-1-AP-150UL
Description
The CAP2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human CAP2. This antibody recognizes human, mouse, rat antigen. The CAP2 antibody has been validated for the following applications: IF, IHC, IP, WB, ELISA analysis.
Specifications
CAP2 | |
Polyclonal | |
Unconjugated | |
150 μL | |
10486 | |
IgG |
Western Blot, Immunoprecipitation, Immunohistochemistry, Immunofluorescence | |
187 μg/mL | |
Rabbit | |
Primary | |
Human, Rat, Mouse |