Learn More
CAPN3 Mouse anti-Human, Unconjugated, Clone: 1G12A5, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67366-1-IG-20UL
Description
The CAPN3 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CAPN3. This antibody recognizes human, pig, mouse, rat antigen. The CAPN3 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
CAPN3 | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human, Mouse, Rat, Pig |
Western Blot, Immunohistochemistry, ELISA | |
1G12A5 | |
Unconjugated | |
20 μL | |
825 | |
IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.