missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ CaptureSelect™ Biotin Anti-C-tag Conjugate

Thermo Scientific™ CaptureSelect™ Biotin Anti-C-tag Conjugate consists of a 13 kDa recombinant single domain antibody fragment (VHH affinity ligand) with high affinity and selectivity for the 4-amino-acid 'C-tag' peptide tag E‐P‐E‐A (glutamic acid - proline - glutamic acid - alanine).

4660.00 SEK - 16375.00 SEK

Specifications

Content And Storage Store at -5°C to -30°C.
Form Liquid
Product Line CaptureSelect™
Type Conjugate
Label or Dye Biotin & Other Haptens
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18476668
View Documents
Thermo Scientific™
7103252100
100 μg
4660.00 SEK
100µg
Please to purchase this item. Need a web account? Register with us today!
18476808
View Documents
Thermo Scientific™
7103252500
500 μg
16375.00 SEK
500µg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CaptureSelect Biotin Anti-C-tag Conjugate consists of a 13 kDa recombinant single domain antibody fragment (VHH affinity ligand) with high affinity and selectivity for the 4-amino-acid 'C-tag' peptide tag E‐P‐E‐A (glutamic acid - proline - glutamic acid - alanine). The Biotin Anti-C-tag affinity ligand recognizes the E-P-E-A peptide when this tag is fused directly to the C-terminus of a protein. The affinity ligand, constructed as a mono-specific bihead, is chemically conjugated to biotin via an appropriate spacer that retains the binding reactivity of the ligand when used in combination with streptavidin-based conjugates or streptavidin pre-coated surfaces.

The CaptureSelect Biotin Anti-C-tag Conjugate allows you to

  • Detect, quantitate, and characterize any C-tag fusion protein

Applications for CaptureSelect Biotin Anti-C-tag Conjugate include ELISA, Western blot, Gyrolab™-based immunoassays, and label-free detection platforms such as those based on surface plasmon resonance (Biacore™ and IBIS-MX96 systems) and bio-layer interferometry (ForteBio™ Octet™ systems).

Animal origin statement

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12–15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

Specifications

Specifications

Store at -5°C to -30°C.
CaptureSelect™
Biotin & Other Haptens
Liquid
Conjugate
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ CaptureSelect™ Biotin Anti-C-tag Conjugate

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.