Learn More
Caspase 2/p18 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 10436-1-AP-20UL
 View more versions of this product
Product Code. 16886823
  • 1645.54 SEK / 20µL
This item is not returnable. View return policy
Description
Description
This gene encodes a protein which is a member of the cysteine-aspartic acid protease (caspase) family. Sequential activation of caspases plays a central role in the execution-phase of cell apoptosis. Caspases exist as inactive proenzymes which undergo proteolytic processing at conserved aspartic residues to produce two subunits, large and small, that dimerize to form the active enzyme. The proteolytic cleavage of this protein is induced by a variety of apoptotic stimuli. Alternative splicing of this gene results in multiple transcript variants that encode different isoforms.
Specifications
Specifications
Caspase 2/p18
Polyclonal
Unconjugated
CASP2
CASP 2, CASP2, Caspase 2, Caspase 2/p18, Caspase2, ICH 1L, ICH 1L/1S, ICH1, NEDD 2, NEDD2, Protease ICH 1
Rabbit
Antigen Affinity Chromatography
RUO
12366, 64314, 835
-20°C
Liquid
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.4 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P29594, P42575, P55215
Casp2
Caspase 2/p18 Fusion Protein Ag0694
20 ÎĽL
Primary
Mouse, Rat, Human
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Caspase 2/p18 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 ÎĽL; Unconjugated