missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Caspase 9/p35/p10 Mouse anti-Human, Mouse, Clone: 1B7G2, Proteintech
Mouse Monoclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | Caspase 9/p35/p10 |
---|---|
Clone | 1B7G2 |
Concentration | 1.5 mg/mL |
Applications | Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16897795
|
Proteintech
66169-1-IG-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16887795
|
Proteintech
66169-1-IG-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Involved in the activation cascade of caspases responsible for apoptosis execution. Binding of caspase-9 to Apaf-1 leads to activation of the protease which then cleaves and activates caspase-3. Promotes DNA damage-induced apoptosis in a ABL1/c-Abl-dependent manner. Proteolytically cleaves poly(ADP-ribose) polymerase (PARP).Specifications
Caspase 9/p35/p10 | |
1.5 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
APAF 3, APAF3, Apoptotic protease Mch 6, CASP 9, CASP9, Caspase 9, Caspase 9/p35/p10, CASPASE 9c, Caspase9, Caspase-9, ICE LAP6, ICE like apoptotic protease 6, MCH6 | |
CASP9 | |
IgG2b | |
Protein A | |
Antibody |
1B7G2 | |
Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
Unconjugated | |
Mouse | |
Mouse, Human | |
P55211, Q8C3Q9 | |
12371, 842 | |
Caspase 9/p35/p10 Fusion Protein Ag20813 | |
Primary | |
-20°C | |
CASP9 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Caspase 9/p35/p10 Mouse anti-Human, Mouse, Clone: 1B7G2, Proteintech