missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Catenin, gamma Mouse, Clone: CTNG/1664, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 3728-MSM4-P0
Denna artikel kan inte returneras.
Se returpolicy
Specifications
| Catenin, gamma | |
| Cell Surface and Cytoplasmic | |
| CTNG/1664 | |
| Unconjugated | |
| NP_001339702 | |
| Mouse | |
| 80-87kDa | |
| Protein A/G Purified | |
| RUO | |
| 3728 | |
| IgG1 κ |
| Immunohistochemistry | |
| Monoclonal | |
| 200μg/mL | |
| 10mM PBS with 0.05% BSA and 0.05% Azide | |
| JUP | |
| Recombinant human full-length gamma catenin protein | |
| A431, T47D or MCF-7 cells. Skin | |
| 20 μg | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction