missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CBLL1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21179-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes an E3 ubiquitin-ligase for the E-cadherin complex and mediates its ubiquitination, endocytosis, and degradation in the lysosomes. The encoded protein contains a RING-finger domain and is also thought to have a role in control of cell proliferation. A related pseudogene has been identified on chromosome X. Alternative splicing results in a non-coding transcript variant.Specifications
CBLL1 | |
Polyclonal | |
Unconjugated | |
CBLL1 | |
c Cbl like protein 1, CBLL1, HAKAI, RING finger protein 188, RNF188 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
104836, 314028, 79872 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.19 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q75N03, Q9JIY2 | |
Cbll1 | |
CBLL1 Fusion Protein Ag15463 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CBLL1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction