Learn More
CBX4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 18544-1-AP-20UL
 View more versions of this product
Product Code. 16878534
  • 1645.54 SEK / 20µL
This item is not returnable. View return policy
Description
Description
Polycomb group (PcG) proteins are chromatin-associated proteins which are important for embryonic and adult stem cell self-renewal and maintece. At least two distinct human PcG complexes have been identified. Polycomb repressive complex I (PRC1) contains CBX4/Pc2, HPH1 and RING domain-containing proteins (RING1, RING2 and BMI1). CBX4 (Chromobox protein homolog 4 or PC2), which functions as a transcriptional suppressor, is a candidate of KyoT2-binding proteins. It is a E3 SUMO-protein ligase which facilitates SUMO1 conjugation. Depletion of CBX4 results in decreased cellular resistance to ionizing radiation. CBX4 is directly involved in the cellular response to DNA damage.
Specifications
Specifications
CBX4
Polyclonal
Unconjugated
CBX4
CBX4, Chromobox protein homolog 4, E3 SUMO protein ligase CBX4, hPC2, NBP16, PC2, Polycomb 2 homolog
Rabbit
Antigen Affinity Chromatography
RUO
12418, 501403, 8535
-20°C
Liquid
Immunocytochemistry, Western Blot, Immunofluorescence
0.27 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O00257, O55187
Cbx4
CBX4 Fusion Protein Ag13425
20 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CBX4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated