Learn More
CCDC105 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 24026-1-AP-20UL
 View more versions of this product
Product Code. 16882615
  • 1645.54 SEK / 20µL
This item is not returnable. View return policy
Specifications
CCDC105
Polyclonal
Unconjugated
CCDC105
CCDC105
Rabbit
Antigen Affinity Chromatography
RUO
126402
-20°C
Liquid
Immunofluorescence, Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin)
0.52 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8IYK2
CCDC105
CCDC105 Fusion Protein Ag21208
20 μL
Primary
Human
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CCDC105 Rabbit anti-Human, Polyclonal, Proteintech > 20 μL; Unconjugated