Learn More
CCDC105 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 24026-1-AP-20UL
 View more versions of this product
Product Code. 16882615
  • 1645.54 SEK / 20µL
Specifications
CCDC105
Polyclonal
Unconjugated
CCDC105
CCDC105
Rabbit
Antigen Affinity Chromatography
RUO
126402
-20°C
Liquid
Immunofluorescence, Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin)
0.52 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8IYK2
CCDC105
CCDC105 Fusion Protein Ag21208
20 μL
Primary
Human
Antibody
IgG
Certificates
Promotions
Promotions
Product Content Correction
Product Title
CCDC105 Rabbit anti-Human, Polyclonal, Proteintech > 20 μL; Unconjugated