missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCDC137 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | CCDC137 |
---|---|
Concentration | 0.3 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16825475
|
Proteintech
27201-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 14-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16815475
|
Proteintech
27201-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 14-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
CCDC137 is also known as the retinoic acid resistance factor protein.Specifications
CCDC137 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q6PK04 | |
339230 | |
CCDC137 Fusion Protein Ag23942 | |
Primary | |
-20°C | |
CCDC137 |
0.3 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
CCDC137 | |
CCDC137 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |