Learn More
CCDC6 Mouse anti-Human, Unconjugated, Clone: 1D6A8, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67637-1-IG-150UL
 View more versions of this product
Product Code. 16870376
  • 4652.90 SEK / 150µL
Description
The CCDC6 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CCDC6. This antibody recognizes human, mouse, rat antigen. The CCDC6 antibody has been validated for the following applications: IF, WB, ELISA analysis.
Specifications
Specifications
CCDC6
Monoclonal
500 μg/mL
Mouse
Primary
Human, Rat, Mouse
Western Blot, Immunofluorescence, ELISA
1D6A8
Unconjugated
150 μL
8030
IgG1
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
CCDC6 Mouse anti-Human, Unconjugated, Clone: 1D6A8, Proteintech > 150 μL