Learn More
CCDC6 Mouse anti-Human, Unconjugated, Clone: 1D6A8, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67637-1-IG-150UL
Description
The CCDC6 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CCDC6. This antibody recognizes human, mouse, rat antigen. The CCDC6 antibody has been validated for the following applications: IF, WB, ELISA analysis.
Specifications
CCDC6 | |
Monoclonal | |
500 μg/mL | |
Mouse | |
Primary | |
Human, Rat, Mouse |
Western Blot, Immunofluorescence, ELISA | |
1D6A8 | |
Unconjugated | |
150 μL | |
8030 | |
IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.