Learn More
CCDC6 Mouse anti-Human, Unconjugated, Clone: 1D6A8, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67637-1-IG-20UL
 View more versions of this product
Product Code. 16880376
  • 1645.54 SEK / 20µL
Description
The CCDC6 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CCDC6. This antibody recognizes human, mouse, rat antigen. The CCDC6 antibody has been validated for the following applications: IF, WB, ELISA analysis.
Specifications
Specifications
CCDC6
Monoclonal
500 μg/mL
Mouse
Primary
Human, Rat, Mouse
Western Blot, Immunofluorescence, ELISA
1D6A8
Unconjugated
20 μL
8030
IgG1
Certificates
Promotions
Promotions
Product Content Correction
Product Title
CCDC6 Mouse anti-Human, Unconjugated, Clone: 1D6A8, Proteintech > 20 μL