missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCL2/JE/MCP-1 Antibody (123616R), PE/Cy7, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB479RPECY7
This item is not returnable.
View return policy
Description
CCL2/JE/MCP-1 Monoclonal antibody specifically detects CCL2/JE/MCP-1 in Mouse samples. It is validated for Flow Cytometry
Specifications
CCL2/JE/MCP-1 | |
Monoclonal | |
PE-Cyanine7 | |
PBS | |
Rat | |
Protein A or G purified from cell culture supernatant | |
RUO | |
6347 | |
Store at 4C in the dark. Do not freeze. | |
IgG2b |
Flow Cytometry | |
123616R | |
Flow Cytometry | |
C-C motif chemokine ligand 2, CCL2, GDCF-2, HC11, HSMCR30, MCAF, Mcp1, Mcp-1, SCYA2, SMC-CF | |
E. coli-derived recombinant mouse CCL2, Gln24-Arg96, Accession # P10148 | |
0.1 mL | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CCL2/JE/MCP-1 Antibody (123616R), PE/Cy7, Novus Biologicals™ > 0.1 mL; PE-Cyanine7
Spot an opportunity for improvement?Share a Content Correction