missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCNYL1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26970-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
CCNYL1 belongs to the cyclin family, Cyclin Y subfamily and contains 1 cyclin N terminal domain. The specific function of CCNYL1 is not yet known.Specifications
CCNYL1 | |
Polyclonal | |
Unconjugated | |
CCNYL1 | |
CCNYL1, cyclin Y like 1, Cyclin Y like protein 1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
151195 | |
-20°C | |
Liquid |
Western Blot | |
0.43 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8N7R7 | |
CCNYL1 | |
CCNYL1 Fusion Protein Ag25583 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |