missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCNYL1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | CCNYL1 |
---|---|
Concentration | 0.43 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16885285
|
Proteintech
26970-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16875285
|
Proteintech
26970-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
CCNYL1 belongs to the cyclin family, Cyclin Y subfamily and contains 1 cyclin N terminal domain. The specific function of CCNYL1 is not yet known.Specifications
CCNYL1 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q8N7R7 | |
151195 | |
CCNYL1 Fusion Protein Ag25583 | |
Primary | |
-20°C | |
CCNYL1 |
0.43 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
CCNYL1, cyclin Y like 1, Cyclin Y like protein 1 | |
CCNYL1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |