missing translation for 'onlineSavingsMsg'
Learn More

CCNYL1 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 26970-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16875285

  • 4721.67 SEK / 150µL
Estimated Shipment: 17-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

CCNYL1 belongs to the cyclin family, Cyclin Y subfamily and contains 1 cyclin N terminal domain. The specific function of CCNYL1 is not yet known.
Specifications

Specifications

CCNYL1
Polyclonal
Unconjugated
CCNYL1
CCNYL1, cyclin Y like 1, Cyclin Y like protein 1
Rabbit
Antigen Affinity Chromatography
RUO
151195
-20°C
Liquid
Western Blot
0.43 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q8N7R7
CCNYL1
CCNYL1 Fusion Protein Ag25583
150 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers