Learn More
CCR10 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 22071-1-AP-20UL
 View more versions of this product
Product Code. 16881615
  • 1645.54 SEK / 20µL
This item is not returnable. View return policy
Description
Description
Chemokines are a group of small (approximately 8 to 14 kDa), mostly basic, structurally related molecules that regulate cell trafficking of various types of leukocytes through interactions with a subset of 7-transmembrane, G protein-coupled receptors. Chemokines also play fundamental roles in the development, homeostasis, and function of the immune system, and they have effects on cells of the central nervous system as well as on endothelial cells involved in angiogenesis or angiostasis. Chemokines are divided into 2 major subfamilies, CXC and CC, based on the arrangement of the first 2 of the 4 conserved cysteine residues; the 2 cysteines are separated by a single amino acid in CXC chemokines and are adjacent in CC chemokines. CCR10 is the receptor for CCL27; CCR10-CCL27 interactions are involved in T cell-mediated skin inflammation.
Specifications
Specifications
CCR10
Polyclonal
Unconjugated
CCR10
C C chemokine receptor type 10, C C CKR 10, CC CKR 10, CCR 10, CCR10, G protein coupled receptor 2, GPR2
Rabbit
Antigen Affinity Chromatography
RUO
12777, 2826
-20°C
Liquid
Flow Cytometry, Western Blot, Immunohistochemistry (Paraffin)
0.44 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P46092, Q9JL21
CCR10
CCR10 Fusion Protein Ag17212
20 μL
Primary
Mouse, Human
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CCR10 Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 20 μL; Unconjugated