missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCR3 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | CCR3 |
---|---|
Concentration | 0.3 mg/mL |
Applications | Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16891885
|
Proteintech
22351-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16881885
|
Proteintech
22351-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemokines play important roles in inflammation and critical for the recruitment of effector immune cells to sites of infection. Chemokines activate leukocytes by binding to G protein coupled receptors. The ever-growing chemokine receptor subtypes can be divided into 2 major groups, CXCR and CCR, based on the 2 major classes of chemokines. One of the CCR receptors, CCR3 (eotaxin receptor), is expressed on eosinophils and certain T cell population respond to a variety of CC chemokines apart from eotaxin, including RANTES, monocyte chemotactic protein (MCP)-2, MCP-3, and MCP-4. CCR3 facilitated infection by a more restricted subset of primary viruses, and binding of the CCR3 ligand, eotaxin, and inhibited infection by these isolates.Specifications
CCR3 | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Human | |
P51677 | |
1232 | |
CCR3 Fusion Protein Ag17926 | |
Primary | |
-20°C | |
CCR3 |
0.3 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
C C chemokine receptor type 3, C C CKR 3, CC CKR 3, CCR 3, CCR3, CD193, CKR3, CMKBR3, Eosinophil eotaxin receptor | |
CCR3 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CCR3 Rabbit anti-Human, Polyclonal, Proteintech