missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCR5 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | CCR5 |
---|---|
Concentration | 387 ÎĽg/mL |
Applications | Immunohistochemistry, Immunofluorescence, Flow Cytometry, ELISA |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16897364
|
Proteintech
17476-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16887364
|
Proteintech
17476-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The CCR5 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human CCR5. This antibody recognizes human antigen. The CCR5 antibody has been validated for the following applications: FC, IHC, ELISA analysis.
Specifications
CCR5 | |
Immunohistochemistry, Immunofluorescence, Flow Cytometry, ELISA | |
Unconjugated | |
Human | |
IgG |
387 ÎĽg/mL | |
Polyclonal | |
Rabbit | |
1234 | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CCR5 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech