missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
CCR8 (CD198) Mouse anti-Human , APC, Clone: 433H, BD Biosciences™
Mouse Monoclonal Antibody
Brand: BD Biosciences 566897
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
CCR8 (CD198) Monoclonal antibody specifically detects CCR8 (CD198) in Human samples. It is validated for Flow Cytometry
Specifikationer
| CCR8 (CD198) | |
| Monoclonal | |
| APC | |
| CCR8; CKR-L1; CKRL1; CMKBR8; CMKBRL2; CY6; GPR-CY6; GPRCY6; MGC129966; MGC1 | |
| Human CCR8 Transfected Cell Line | |
| RUO | |
| Human | |
| IgG2a κ |
| Flow Cytometry | |
| 433H | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Mouse | |
| 100 Tests | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to APC under optimum conditions, and unconjugated antibody and free APC were removed. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction