Learn More
CCRL2 Mouse anti-Human, Clone: 2C1E11, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 66611-1-IG-20UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16888645
  • 1355.00 SEK / 20µL
Estimated Shipment: 23-03-2023
to see stock.
Description
Description
CCRL2 (C-C chemokine receptor-like 2), also known as chemokine receptor X, human chemokine receptor (HCR), CRAM-A or CRAM-B, is a seven pass transmembrane protein expressed by monocytes, neutrophils, and dendritic cells of immunal tissues (predomily spleen, fetal liver, bone marrow and lymph node). It shares over 40% homology with other C-C chemokine receptors but does not share the conserved DRY motif which is among the important motifs necessary for signalling and ligand-binding. C-C chemokine receptors are G-protein coupled, seven pass transmembrane domain proteins whose major physiological role is to function in the chemotaxis of T cells and phagocytic cells to areas of inflammation.
Specifications
Specifications
CCRL2
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O00421
CCRL2
CCRL2 Fusion Protein Ag4045
20 μL
Primary
Human
Antibody
IgG1
Western Blot, Immunohistochemistry (Paraffin)
2C1E11
Unconjugated
CCRL2
C C chemokine receptor like 2, CCR11, CCR6, CCRL2, Chemokine receptor CCR11, Chemokine receptor X, CKRX, CRAM, CRAM A, CRAM B, HCR
Mouse
Protein G
RUO
9034
-20°C