Learn More
CCT8 Mouse anti-Human, Unconjugated, Clone: 1A7C3, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67539-1-IG-20UL
Description
The CCT8 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CCT8. This antibody recognizes human, mouse, rat antigen. The CCT8 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
CCT8 | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human, Rat, Mouse |
Western Blot, Immunohistochemistry, ELISA | |
1A7C3 | |
Unconjugated | |
20 μL | |
10694 | |
IgG2a |