missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD13 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14553-1-AP-20UL
This item is not returnable.
View return policy
Description
CD13 (ANPEP) is a 150-170 kDa type II transmembrane zinc-binding ectopeptidase expressed on various cell types. CD13 is a metalloprotease that preferentially catalyzes removal of neutral amino acids from small peptides, thus activating or inactivating bioactive peptides. CD13 also has a role in extracellular matrix degradation, antigen processing and signal transduction, is important in inflammatory responses, regulates intercellular contact, cell motility and vascularization. CD13 is expressed by granulocytes, monocytes, myeloid leukemia cells, and is involved in protection of leukemic cells against apoptosis and its expression associated with poor prognosis of carcinomas. CD13 functions as an aminopeptidase enzyme and is also alpha receptor for coronavirus.Specifications
CD13 | |
Polyclonal | |
Unconjugated | |
ANPEP | |
Alanyl aminopeptidase, Aminopeptidase M, Aminopeptidase N, ANPEP, AP M, AP N, APN, CD13, gp150, hAPN, LAP1, Microsomal aminopeptidase, p150, PEPN | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
16790, 290, 81641 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Flow Cytometry | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P15144, P15684, P97449 | |
ANPEP | |
CD13 Fusion Protein Ag5976 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CD13 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction