Learn More
CD133-1,2,4,7 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 19945-1-AP-150UL
 View more versions of this product
Product Code. 16889414
  • 4652.90 SEK / 150µL
This item is not returnable. View return policy
Description
Description
The CD133-1, 2, 4, 7 antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human CD133-1, 2, 4, 7. This antibody recognizes human, mouse antigen. The CD133-1, 2, 4, 7 antibody has been validated for the following applications: IHC, IP, WB, ELISA analysis.
Specifications
Specifications
CD133-1,2,4,7
Polyclonal
Unconjugated
150 ÎĽL
8842
IgG
Western Blot, Immunoprecipitation, Immunohistochemistry, ELISA
167 ÎĽg/mL
Rabbit
Primary
Human, Mouse
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CD133-1,2,4,7 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech > 150 ÎĽL