Learn More
CD138 (Syndecan-1) Rat, anti-Mouse, Biotin, Clone: 281-2, STEMCELL Technologies
Rat Monoclonal Antibody
Brand:  Stemcell Technologies 60035BT
Encompass_Preferred
 View more versions of this product
Product Code. 17158671
  • 7209.00 SEK / 500µg
Description
  • The 281-2 antibody reacts with an extracellular epitope on the core protein of mouse CD138 (Syndecan-1), a ∼60 - 100 kDa type 1 transmembrane glycoprotein expressed on the surface of pre-B cells, immature B cells, and normal and malignant plasma cells (but not mature circulating B cells)
  • CD138 is also expressed on non-hematopoietic cells such as embryonic mesenchymal cells, endothelial, epithelial and neural cells
  • CD138 is thought to act primarily as a receptor that modulates cell proliferation, cell migration and cell-matrix associations by linking the extracellular matrix to the cytoskeleton
  • Theparin sulfate and chondroitin sulfate moieties attached to CD138 associate with several proteins, including collagens, fibronectin, tenascin, thrombospondin and certain cytokines
Specifications
Specifications
CD138 (Syndecan-1)
Monoclonal
Biotin
500 μg
Mouse
B Cells, Plasma
Immunohistochemistry, Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence
281-2
Rat
Primary
IgG2a κ
Certificates
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
CD138 (Syndecan-1) Rat, anti-Mouse, Biotin, Clone: 281-2, STEMCELL Technologies