missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD1d Mouse anti-Rat, BV421, Clone: WTH2 (also known as WTH-2), BD OptiBuild™
Mouse Monoclonal Antibody
Brand: BD Biosciences 744188
4440.50 SEK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
CD1d Monoclonal antibody specifically detects CD1d in Rat samples. It is validated for Flow Cytometry.
Specifications
CD1d | |
Monoclonal | |
0.2 mg/mL | |
Mouse | |
RUO | |
Rat | |
Purified |
Flow Cytometry | |
WTH2 (also known as WTH-2) | |
Brilliant Violet 421 | |
50 ÎĽg | |
Primary | |
Refrigerate (2 to 8°C) | |
IgG2a Îş |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CD1d Mouse anti-Rat, BV421, Clone: WTH2 (also known as WTH-2), BD OptiBuild™ > Rat CD1d BV421 WTH2 50 μg
Spot an opportunity for improvement?Share a Content Correction