missing translation for 'onlineSavingsMsg'
Learn More

CD226 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 17842-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16887834

  • 3665.00 SEK / 150µL
Estimated Shipment: 27-03-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a glycoprotein expressed on the surface of NK cells, platelets, monocytes and a subset of T cells. It is a member of the Ig-superfamily containing 2 Ig-like domains of the V-set. The protein mediates cellular adhesion of platelets and megakaryocytic cells to vascular endothelial cells. The protein also plays a role in megakaryocytic cell maturation.
Specifications

Specifications

CD226
Polyclonal
Unconjugated
CD226
CD226, CD226 antigen, CD226 molecule, DNAM 1, DNAM1, DNAX accessory molecule 1, PTA1, TLiSA1
Rabbit
Antigen Affinity Chromatography
RUO
10666
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin)
0.21 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q15762
CD226
CD226 Fusion Protein Ag12206
150 μL
Primary
Human
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers