Läs mer
CD235ab (Glycophorin A/B) Mouse, anti-Human, PE, Clone: HIR2 (GA-R2), STEMCELL Technologies
Mouse Monoclonal Antibody
Brand: Stemcell Technologies 60111PE.1
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
- The HIR2 (GA-R2) antibody recognizes an epitope common to the N-terminal region of human CD235a (glycophorin A) and CD235b (glycophorin B), homologous type I sialoglycoproteins of the erythrocyte (red blood cell) membrane that bear the antigenic determinants for the MN and Ss blood groups
- CD235ab is expressed on early- and late-stage erythroblasts, mature erythrocytes, and erythroid cell lines such as K562 and HEL, but not on other cell types
- CD235a is abundantly expressed whereas CD235b is a relatively minor membrane component
- The proteins are believed to provide a large mucin-like surface to erythrocytes that acts to minimize aggregation in the circulation
- The HIR2 antibody binds with higher affinity to CD235a than CD235b, and agglutinates untreated erythrocytes
Specifikationer
| CD235ab (Glycophorin A/B) | |
| Monoclonal | |
| PE | |
| 25 μg | |
| Human | |
| Red Blood Cells |
| Immunohistochemistry, Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| HIR2 (GA-R2) | |
| Mouse | |
| Primary | |
| IgG2b κ |
Korrigering av produktinnehåll
Fisher Scientific Encompass-programmet erbjuder artiklar som inte ingår i vår distributionsportfölj. Dessa produkter har vanligtvis inga bilder eller detaljerade beskrivningar. Vi är dock angelägna om att förbättra din shoppingupplevelse. Använd formuläret nedan för att ge feedback relaterad till innehållet på denna produkt.