Learn More
CD24 Mouse anti-Human, Unconjugated, Clone: 1H5C4, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67627-1-IG-20UL
 View more versions of this product
Product Code. 16880356
  • 1645.54 SEK / 20µL
Description
The CD24 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CD24. This antibody recognizes human antigen. The CD24 antibody has been validated for the following applications: FC, WB, ELISA analysis.
Specifications
Specifications
CD24
Monoclonal
1000 μg/mL
Mouse
Primary
Human
Western Blot, Flow Cytometry, ELISA
1H5C4
Unconjugated
20 μL
100133941
IgG1
Certificates
Promotions
Promotions
Product Content Correction
Product Title
CD24 Mouse anti-Human, Unconjugated, Clone: 1H5C4, Proteintech > 20 μL