missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD24 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1706.43 SEK - 4863.32 SEK
Specifications
Antigen | CD24 |
---|---|
Concentration | 0.18 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
CD24 is a two-chain glycosylphosphatidylinositol (GPI)-anchored glycoprotein expressed at multiple stages of B-cell development, beginning with the bone marrow pro-B-cell compartment and continuing through mature, surface Ig positive B-cells. Plasma cell expression is very low or negative. CD24 is also expressed on the majority of B-lineage acute lymphoblastic leukemias, B-cell CCLs and B-cell non-Hodgkin′s lymphomas. CD24 may play a role in regulation of B-cell proliferation and maturation, and control of autoimmunity.Specifications
CD24 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
P24807, P25063 | |
100133941, 12484 | |
CD24 Fusion Protein Ag11679 | |
Primary | |
-20°C | |
CD24 |
0.18 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
CD24, CD24 molecule, CD24A, Signal transducer CD24 | |
CD24 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CD24 Rabbit anti-Human, Mouse, Polyclonal, Proteintech