missing translation for 'onlineSavingsMsg'
Learn More

BD Biosciences CD3 Mouse anti-Human , BB700, Clone: OKT3, BD Biosciences™

Mouse Monoclonal Antibody

Brand:  BD Biosciences 566819

Additional Details : Weight : 0.00900kg

 View more versions of this product

Product Code. 17227634

  • 2485.00 SEK / 25 tests
Estimated Shipment: 26-06-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

CD3 Monoclonal antibody specifically detects CD3 in Human samples. It is validated for Flow Cytometry
Specifications

Specifications

CD3
Monoclonal
BB700
CD3E; CD3e; T-cell surface antigen T3/Leu-4 epsilon; T3E; TCRE
Sheep Erythrocyte Rosette-purified Human T Cells
RUO
Human
Mouse BALB/c x A/J, also known as CAF1 IgG2a κ
Flow Cytometry
OKT3
Aqueous buffered solution containing ≤0.09% sodium azide.
Mouse
25 Tests
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BB700 under optimum conditions, and unconjugated antibody and free BD Horizon BB700 were removed.
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers