Learn More
CD300A Mouse anti-Human, Unconjugated, Clone: 2F9C5, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67242-1-IG-20UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16849765
  • 1656.73 SEK / 20µL
Estimated Shipment: 16-05-2024
to see stock.
Description
Description
The CD300A antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CD300A. This antibody recognizes human antigen. The CD300A antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
CD300A
Monoclonal
1000 μg/mL
Mouse
Primary
Human
Western Blot, ELISA
2F9C5
Unconjugated
20 μL
11314
IgG1