missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD300A Mouse anti-Human, Unconjugated, Clone: 2F9C5, Proteintech
Mouse Monoclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | CD300A |
---|---|
Clone | 2F9C5 |
Concentration | 1000 μg/mL |
Applications | Western Blot, ELISA |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16849765
|
Proteintech
67242-1-IG-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16839765
|
Proteintech
67242-1-IG-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The CD300A antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CD300A. This antibody recognizes human antigen. The CD300A antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
CD300A | |
1000 μg/mL | |
Monoclonal | |
Mouse | |
11314 | |
Primary |
2F9C5 | |
Western Blot, ELISA | |
Unconjugated | |
Human | |
IgG1 |