Learn More
CD300A Mouse anti-Human, Unconjugated, Clone: 2F9C5, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67242-1-IG-150UL
Additional Details : Weight : 0.00010kg
Description
The CD300A antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CD300A. This antibody recognizes human antigen. The CD300A antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
CD300A | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human |
Western Blot, ELISA | |
2F9C5 | |
Unconjugated | |
150 μL | |
11314 | |
IgG1 |