missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD300LF Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13334-1-AP-20UL
This item is not returnable.
View return policy
Description
CLM1 is an inhibitory receptor of the Ig superfamily expressed on myeloid cells. It mediates negative regulatory signals by recruiting SHP1 (PTPN6, MIM 176883) or SHIP.Specifications
CD300LF | |
Polyclonal | |
Unconjugated | |
CD300LF | |
CD300f, CD300LF, CLM 1, CLM1, CMRF35 like molecule 1, IgSF13, IREM 1, IREM1, NK inhibitory receptor, NKIR | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
146722 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.18 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8TDQ1 | |
CD300LF | |
CD300LF Fusion Protein Ag4150 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CD300LF Rabbit anti-Human, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction