missing translation for 'onlineSavingsMsg'
Learn More

CD34 Mouse anti-Human, Clone: 3C8G12, Proteintech

Mouse Monoclonal Antibody

1662.46 SEK - 4700.76 SEK

Specifications

Antigen CD34
Clone 3C8G12
Concentration 1 mg/mL
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16887155
View Documents
Proteintech
60180-1-IG-20UL
20 ÎĽL
1662.46 SEK
20µL
Please to purchase this item. Need a web account? Register with us today!
16877155
View Documents
Proteintech
60180-1-IG-150UL
150 ÎĽL
4700.76 SEK
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CD34 is a highly glycosylated monomeric with a molecular weight range of 111-115 kDa surface protein that is present on many stem cell populations. CD34 is a stem cell marker although its expression on human hematopoietic stem cells is reversible. CD34 may serve as a surface receptor that undergoes receptor-mediated endocytosis and regulates adhesion, differentiation and proliferation of hematopoietic stem cells and other progenitors. CD34 expression is likely to represent a specific state of hematopoietic development that may have altered adhering properties with expanding and differentiating capabilities in both in vitro and in vivo conditions. CD34 is possibly an adhesion molecule with a putative role for mediating the attachment of stem cells to the bone marrow extracellular matrix or directly to stromal cells. Further, CD34 could act as a scaffold for the attachment of lineage specific glycans, allowing stem cells to bind to lectins expressed by stromal cells or other marrow components. CD34 is thought to have a role in presenting carbohydrate ligands to selectins. The intracellular chain of the CD34 antigen is a site of phosphorylation by activated protein kinase C suggesting a putative role in signal transduction. Diseases associated with CD34 dysfunction include dermatofibrosarcoma and neurofibroma.
Specifications

Specifications

CD34
1 mg/mL
Monoclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
CD34, CD34 molecule
CD34
IgG2b
Caprylic Acid Precipitation
Antibody
3C8G12
Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
Mouse
Human
P28906
947
CD34 Fusion Protein Ag5996
Primary
-20°C
CD34
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
CD34 Mouse anti-Human, Clone: 3C8G12, Proteintech

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.