missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD366 (TIM-3) Rat anti-Human, Brilliant Violet 650, Clone: 344823, BD OptiBuild™
Rat Monoclonal Antibody
Brand: BD Biosciences 747960
6019.82 SEK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
CD366 (TIM-3) Monoclonal antibody specifically detects CD366 (TIM-3) in Human samples. It is validated for Flow Cytometry.
Specifications
CD366 (TIM-3) | |
Monoclonal | |
0.2 mg/mL | |
Rat | |
RUO | |
CD366 (TIM-3) | |
Refrigerate (2 to 8°C) |
Flow Cytometry | |
344823 | |
Brilliant Violet 650 | |
50 ÎĽg | |
Primary | |
Human | |
IgG2a Îş |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CD366 (TIM-3) Rat anti-Human, Brilliant Violet 650, Clone: 344823, BD OptiBuild™ > 50 μg; Brilliant Violet 650
Spot an opportunity for improvement?Share a Content Correction