missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD39/ENTPD1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 19229-1-AP-150UL
This item is not returnable.
View return policy
Description
19229-1-AP recognizes both the 3 isoforms and soluble form of CD39 (38-478 aa) molecule.
CD39 cell surface antigen is a 70-100kD molecule expressed on peripheral blood B cells, monocytes, T cell clones, and also weakly expressed on granulocytes. CD39 has intrinsic ecto-ATPase activity. Expression of CD39 is induced on T cells and increased on B cells as a late activation antigen.Specifications
CD39/ENTPD1 | |
Polyclonal | |
Unconjugated | |
ENTPD1 | |
ATPDase, CD39, Ecto apyrase, Ecto ATP diphosphohydrolase 1, Ecto ATPase 1, Ecto ATPDase 1, ENTPD1, NTPDase 1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
12495, 64519, 953 | |
-20°C | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P49961, P55772, P97687 | |
Entpd1 | |
CD39/ENTPD1 Fusion Protein Ag6165 | |
150 ÎĽL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CD39/ENTPD1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 ÎĽL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction