missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD4 Mouse anti-Human , BV510, Clone: OKT4, BD Biosciences™
Mouse Monoclonal Antibody
3700.00 SEK - 7525.00 SEK
Specifications
Antigen | CD4 |
---|---|
Clone | OKT4 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | Brilliant Violet 510 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17217624
|
BD Biosciences
566805 |
25 Tests |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17207624
|
BD Biosciences
566804 |
100 Tests |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
CD4 Monoclonal antibody specifically detects CD4 in Human samples. It is validated for Flow CytometrySpecifications
CD4 | |
Flow Cytometry | |
Brilliant Violet 510 | |
RUO | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Sheep Erythrocyte Rosette-purified Human T Cells | |
Primary |
OKT4 | |
Monoclonal | |
Mouse | |
Human | |
CD4 antigen (p55); T-cell surface antigen T4/Leu-3 | |
IgG2b Kappa | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BV510 under optimum conditions, and unconjugated antibody and free BD Horizon™ BV510 were removed. |