Learn More
CD40 Mouse anti-Human, Unconjugated, Clone: 2A8G5, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 66965-1-IG-20UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16889295
  • 1355.00 SEK / 20µL
Estimated Shipment: 28-03-2023
to see stock.
Description
Description
The CD40 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CD40. This antibody recognizes human antigen. The CD40 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
Specifications
CD40
Monoclonal
1000 μg/mL
Mouse
Primary
Human
ELISA, Immunofluorescence, Immunohistochemistry, Western Blot
2A8G5
Unconjugated
20 μL
958
IgG1