Learn More
Proteintech CD40 Mouse anti-Human, Unconjugated, Clone: 2A8G5, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66965-1-IG-150UL
Additional Details : Weight : 0.00010kg
Description
The CD40 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CD40. This antibody recognizes human antigen. The CD40 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
CD40 | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human |
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
2A8G5 | |
Unconjugated | |
150 μL | |
958 | |
IgG1 |