Learn More
CD42c Rabbit, anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 55161-1-AP-150UL
 View more versions of this product
Product Code. 16886635
  • 4700.76 SEK / 150µL
Description
The CD42c antibody from Proteintech is a rabbit polyclonal antibody to a peptide of human CD42c. This antibody recognizes human antigen. The CD42c antibody has been validated for the following applications: WB,ELISA analysis.
Specifications
Specifications
CD42c
Polyclonal
Unconjugated
150 μL
2812
Liquid
Western Blot, ELISA
180 μg/mL
Rabbit
Primary
Human
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
CD42c Rabbit, anti-Human, Unconjugated, Polyclonal, Proteintech > 150 μL