Learn More
CD42c Rabbit, anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 55161-1-AP-150UL
This item is not returnable.
View return policy
Description
The CD42c antibody from Proteintech is a rabbit polyclonal antibody to a peptide of human CD42c. This antibody recognizes human antigen. The CD42c antibody has been validated for the following applications: WB,ELISA analysis.
Specifications
CD42c | |
Polyclonal | |
Unconjugated | |
150 μL | |
2812 | |
Liquid |
Western Blot, ELISA | |
180 μg/mL | |
Rabbit | |
Primary | |
Human |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CD42c Rabbit, anti-Human, Unconjugated, Polyclonal, Proteintech > 150 μL
Spot an opportunity for improvement?Share a Content Correction